siRNA Details
Gene Symbol: Has1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172323.1 | NP_758826.1 | 2 | 355 | ||
GenBank mRNA | AB097568.1 | - | 2 | 355 |

More Information
Other Species Targeted

Gene Ontology Categories:
