siRNA Details
Gene Symbol: Xlr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229630.2 | XP_006229692.1 | 2 | 770 | ||
XM_008759622.1 | XP_008757844.1 | 2 | 863 |
Gene Symbol: LOC100911047 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229628.2 | XP_006229690.1 | 2 | 771 | ||
XM_008759621.1 | XP_008757843.1 | 2 | 864 |
