siRNA Details
Gene Symbol: Myl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109495.1 | NP_001102965.1 | 2, 3 | 229 | ||
GenBank mRNA | X51531.1 | - | 2, 3 | 208 |

More Information
Other Species Targeted

Gene Ontology Categories:
