siRNA Details
Gene Symbol: Cfhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001068704.5 | XP_001068704.3 | 2 | 257 | ||
XM_006249947.2 | XP_006250009.1 | 2 | 258 | |||
XM_006249948.2 | XP_006250010.1 | 2 | 258 | |||
XM_006249949.2 | XP_006250011.1 | 2 | 257 | |||
XM_006249950.2 | XP_006250012.1 | 2 | 257 | |||
XM_006249951.2 | XP_006250013.1 | 2 | 258 | |||
GenBank mRNA | FQ211198.1 | - | 2 | 237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
