siRNA Details
Gene Symbol: Kif26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240635.2 | XP_006240697.1 | 16 | 6038 | ||
XM_006240636.2 | XP_006240698.1 | 16 | 6035 |

More Information
Other Species Targeted

Gene Ontology Categories:
