siRNA Details
Gene Symbol: Tmc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126301.1 | NP_001119773.1 | 7 | 992 | ||
XM_006247818.2 | XP_006247880.1 | 7 | 1440 | |||
XM_006247819.2 | XP_006247881.1 | 7 | 1437 | |||
XM_006247821.2 | XP_006247883.1 | 7 | 974 | |||
XM_008768392.1 | XP_008766614.1 | 7 | 965 | |||
GenBank mRNA | BC160873.1 | - | 7 | 992 | 7366375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
