siRNA Details
Gene Symbol: Eif4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100158.2 | NP_001093628.1 | 2 | 271 | ||
GenBank mRNA | BC105875.1 | - | 2 | 216 | 7130292 | |
CO559861.1 | - | 2 | 271 |

More Information
Other Species Targeted

Gene Ontology Categories:
