siRNA Details
Gene Symbol: Efemp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277341.1 | NP_001264270.1 | 5 | 568 | ||
XM_006230742.2 | XP_006230804.1 | 5 | 908 | |||
XM_006230743.1 | XP_006230805.1 | 5 | 559 | |||
XM_006230744.2 | XP_006230806.1 | 5 | 565 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
