siRNA Details
Gene Symbol: Ccdc137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143896.1 | NP_001137368.1 | 2 | 265 | ||
XM_008768483.1 | XP_008766705.1 | 2 | 305 | |||
GenBank mRNA | BC158793.1 | - | 2 | 232 |
