siRNA Details
Gene Symbol: Cux2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271380.1 | NP_001258309.1 | 18 | 2939 | ||
XM_008769215.1 | XP_008767437.1 | 13 | 2584 | |||
XM_008769216.1 | XP_008767438.1 | 13 | 2563 | |||
XM_008769217.1 | XP_008767439.1 | 13 | 2551 | |||
XM_008769218.1 | XP_008767440.1 | 13 | 2530 | |||
XM_008769219.1 | XP_008767441.1 | 17 | 4040 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
