siRNA Details
Gene Symbol: Lrrc45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127571.1 | NP_001121043.1 | 4, 5 | 501 | ||
XM_006247924.1 | XP_006247986.1 | 4, 5 | 556 | |||
GenBank mRNA | BC161940.1 | - | 387 | 9023431 |
