siRNA Details
Gene Symbol: Foxk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107075.1 | NP_001100545.1 | 2 | 136 | ||
XM_006247890.2 | XP_006247952.1 | 2 | 533 | |||
XM_006247891.2 | XP_006247953.1 | 2 | 533 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
