siRNA Details
Gene Symbol: Ttn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761978.1 | XP_008760200.1 | 186 | 39721 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011652, NM_028004, AK157782.
Gene Ontology Categories:
