siRNA Details
Gene Symbol: Mab21l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109497.1 | NP_001102967.1 | 1 | 1444 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010750, BC014750, AK032114, AF228913, AK043552, and others.
Gene Ontology Categories:
