siRNA Details
Gene Symbol: Dydc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134797.1 | NP_001128269.1 | 5 | 500 | ||
XM_006252813.2 | XP_006252875.1 | 3 | 481 | |||
XM_006252814.2 | XP_006252876.1 | 4 | 392 | |||
XM_006252816.2 | XP_006252878.1 | 3 | 321 | |||
XM_008771149.1 | XP_008769371.1 | 3 | 344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
