siRNA Details
Gene Symbol: Higd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106102.1 | NP_001099572.1 | 1 | 167 | ||
GenBank mRNA | BC166544.1 | - | 1 | 137 | 7301761 |

More Information
Other Species Targeted

Gene Ontology Categories:
