siRNA Details
Gene Symbol: Tgfbr3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001067872.5 | XP_001067872.3 | 3 | 556 | ||
XM_006248789.2 | XP_006248851.1 | 2 | 762 | |||
XM_006248790.2 | XP_006248852.1 | 2 | 762 | |||
XM_008769017.1 | XP_008767239.1 | 2 | 762 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
