siRNA Details
Gene Symbol: Slirp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109507.1 | NP_001102977.1 | 3 | 349 | ||
GenBank mRNA | BC166913.1 | - | 3 | 260 | ||
FQ211825.1 | - | 3 | 260 | |||
FQ223545.1 | - | 3 | 260 |

More Information
Other Species Targeted

Gene Ontology Categories:
