siRNA Details
Gene Symbol: Slirp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109507.1 | NP_001102977.1 | 1, 2 | 194 | ||
GenBank mRNA | BC166913.1 | - | 1, 2 | 105 | ||
FQ223545.1 | - | 1, 2 | 105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
