siRNA Details
Gene Symbol: Slirp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109507.1 | NP_001102977.1 | 3 | 340 | ||
GenBank mRNA | BC166913.1 | - | 3 | 251 | ||
FQ211825.1 | - | 3 | 251 | |||
FQ223545.1 | - | 3 | 251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
