siRNA Details
Gene Symbol: Trim8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128083.1 | NP_001121555.1 | 1 | 1076 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_053100, AK155303, BC152344, BC152343, BC029283, and others.
Gene Ontology Categories:
