siRNA Details
Gene Symbol: Nap1l5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044293.1 | NP_001037758.1 | 1 | 370 | ||
XM_006236587.2 | XP_006236649.1 | 1 | 1920 | |||
GenBank mRNA | BC087702.1 | - | 1 | 370 | 7313964 | |
FQ212618.1 | - | 1 | 361 |
