siRNA Details
Gene Symbol: Dtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107157.1 | NP_001100627.1 | 10 | 1961 | ||
XM_006249158.1 | XP_006249220.1 | 9 | 1907 | |||
XM_006249159.1 | XP_006249221.1 | 8 | 1769 | |||
XM_008769137.1 | XP_008767359.1 | 9 | 1762 | |||
GenBank mRNA | AY724523.1 | - | 7 | 1604 | ||
BC166738.1 | - | 9 | 1848 | 7937292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
