siRNA Details
Gene Symbol: Mef2bnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145726.1 | NP_001139198.1 | 6 | 464 | ||
GenBank mRNA | CB782819.1 | - | 6 | 484 | ||
DV714338.1 | - | 6 | 447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
