siRNA Details
Gene Symbol: Cyb5rl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001069267.4 | XP_001069267.2 | 3, 4 | 520 | ||
XM_006238551.2 | XP_006238613.1 | 3, 4 | 522 | |||
XM_008763922.1 | XP_008762144.1 | 3, 4 | 509 | |||
XM_008763923.1 | XP_008762145.1 | 3, 4 | 510 | |||
XM_008763924.1 | XP_008762146.1 | 3, 4 | 508 | |||
XM_008763925.1 | XP_008762147.1 | 3, 4 | 496 | |||
XM_008763926.1 | XP_008762148.1 | 3, 4 | 500 | |||
XM_008763927.1 | XP_008762149.1 | 3, 4 | 504 | |||
XM_008763929.1 | XP_008762151.1 | 3, 4 | 521 | |||
XM_008763930.1 | XP_008762152.1 | 3, 4 | 620 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
