siRNA Details
Gene Symbol: Parvg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130583.1 | NP_001124055.1 | 9 | 734 | ||
XM_006241411.1 | XP_006241473.1 | 9 | 691 | |||
XM_006241412.2 | XP_006241474.1 | 9 | 944 |

More Information
Other Species Targeted

Gene Ontology Categories:
