siRNA Details
Gene Symbol: Snx29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109526.2 | NP_001102996.2 | 16 | 1857 | ||
XM_006245736.2 | XP_006245798.1 | 15 | 2163 | |||
XM_008767497.1 | XP_008765719.1 | 16 | 2972 | |||
XM_008767498.1 | XP_008765720.1 | 14 | 1798 | |||
XM_008767499.1 | XP_008765721.1 | 9 | 3375 |
