siRNA Details
Gene Symbol: LOC690806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135576.2 | NP_001129048.1 | 195 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001135577, AK082625, BC066054, BC094222, BC024659.
