siRNA Details
Gene Symbol: Gabarapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044294.1 | NP_001037759.1 | 2 | 316 | ||
GenBank mRNA | BC105627.1 | - | 2 | 316 | 7323087 | |
FQ213425.1 | - | 2 | 228 |

More Information
Other Species Targeted

Gene Ontology Categories:
