siRNA Details
Gene Symbol: SGSM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014853.2 | NP_055668.2 | 11, 12 | 1462 | ||
XM_011524101.2 | XP_011522403.1 | 12, 13 | 1538 | |||
XM_011524102.2 | XP_011522404.1 | 12, 13 | 1536 | |||
XM_011524103.2 | XP_011522405.1 | 12, 13 | 1537 | |||
XM_011524104.2 | XP_011522406.1 | 12, 13 | 2062 | |||
XM_011524106.1 | XP_011522408.1 | 11, 12 | 1360 | |||
XM_011524107.2 | XP_011522409.1 | 12, 13 | 1539 | |||
XM_017025476.1 | XP_016880965.1 | 6, 7 | 697 | |||
GenBank mRNA | AB275762.1 | - | 11, 12 | 1297 | ||
BC039204.1 | - | 11, 12 | 1462 | 4817100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
