siRNA Details
Gene Symbol: Spdef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109530.1 | NP_001103000.1 | 2 | 556 | ||
XM_006256229.2 | XP_006256291.1 | 2 | 1316 | |||
XM_008772827.1 | XP_008771049.1 | 2 | 426 | |||
GenBank mRNA | BC166744.1 | - | 2 | 560 | 7938752 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
