siRNA Details
Gene Symbol: Tpd52l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044295.1 | NP_001037760.1 | 3 | 271 | ||
XM_006227737.2 | XP_006227799.1 | 3 | 364 | |||
XM_006227739.2 | XP_006227801.1 | 3 | 364 | |||
XM_008758674.1 | XP_008756896.1 | 3 | 363 | |||
GenBank mRNA | BC099809.1 | - | 3 | 271 | 5621894 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
