siRNA Details
Gene Symbol: Amtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044296.1 | NP_001037761.1 | 3, 4 | 239 | ||
XM_008770057.1 | XP_008768279.1 | 2, 3 | 1895 | |||
GenBank mRNA | AM231717.1 | - | 3, 4 | 199 | ||
DQ198381.1 | - | 3, 4 | 239 |

More Information
Other Species Targeted

Gene Ontology Categories:
