siRNA Details
Gene Symbol: Dram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025018.1 | NP_001020189.1 | 6 | 569 | ||
XM_006233120.2 | XP_006233182.1 | 6 | 513 | |||
XM_006233121.2 | XP_006233183.1 | 5 | 453 | |||
XM_006233122.2 | XP_006233184.1 | 5 | 554 | |||
XM_006233123.2 | XP_006233185.1 | 6 | 515 | |||
GenBank mRNA | BC091251.1 | - | 6 | 569 | 7372838 | |
FQ218349.1 | - | 6 | 578 | |||
FQ228782.1 | - | 6 | 666 |
Gene Symbol: LOC689412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109538.1 | NP_001103008.1 | 1 | 216 |
