siRNA Details
Gene Symbol: Sntb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168674.1 | NP_001162145.1 | 2 | 627 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009229, BC145442, AK165009, AK080059, AK040423, and others.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
