siRNA Details
Gene Symbol: LOC498711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771553.1 | XP_008769775.1 | 4 | 2259 | ||
XM_008771554.1 | XP_008769776.1 | 3 | 2211 |
Gene Symbol: LOC102547069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771530.1 | XP_008769752.1 | 1 | 218 |
