siRNA Details
Gene Symbol: Spink13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109539.1 | NP_001103009.1 | 4, 5 | 428 | ||
XM_008772175.1 | XP_008770397.1 | 4, 5 | 350 |

More Information
Other Species Targeted

Gene Ontology Categories:
