siRNA Details
Gene Symbol: LOC689587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253734.2 | XP_006253796.1 | 4 | 2274 | ||
XM_006253735.2 | XP_006253797.1 | 4 | 2253 | |||
XM_008771555.1 | XP_008769777.1 | 3 | 1775 |
