siRNA Details
Gene Symbol: LOC100911565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767440.1 | XP_008765662.1 | 3 | 1498 |
Gene Symbol: LOC689599 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750754.3 | XP_003750802.1 | 4 | 1715 |
