siRNA Details
Gene Symbol: Adam23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767089.1 | XP_008765311.1 | 26 | 5660 | ||
XM_008767090.1 | XP_008765312.1 | 25 | 5569 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adam23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767089.1 | XP_008765311.1 | 26 | 5660 | ||
XM_008767090.1 | XP_008765312.1 | 25 | 5569 |