siRNA Details
Gene Symbol: Cdc42ep4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107063.2 | NP_001100533.1 | 2 | 607 | ||
XM_006247688.2 | XP_006247750.1 | 2 | 1410 | |||
XM_006247689.2 | XP_006247751.1 | 2 | 586 | |||
XM_006247690.2 | XP_006247752.1 | 2 | 1996 | |||
GenBank mRNA | BC161986.1 | - | 2 | 607 | 9030824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
