siRNA Details
Gene Symbol: LOC689757 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101019.2 | NP_001094489.2 | 3 | 344 | ||
XM_008763332.1 | XP_008761554.1 | 3 | 647 | |||
GenBank mRNA | EF100687.1 | - | 3 | 314 | ||
FJ404472.1 | - | 3 | 344 |
Gene Symbol: LOC689770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142304.1 | NP_001135776.1 | 314 | |||
GenBank mRNA | EU128747.1 | - | 320 | |||
FJ404474.1 | - | 314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
