siRNA Details
Gene Symbol: LOC689766 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109549.1 | NP_001103019.1 | 4, 5 | 495 | ||
XM_006250332.2 | XP_006250394.1 | 4, 5 | 530 | |||
XM_008769801.1 | XP_008768023.1 | 2, 3 | 621 |
