siRNA Details
Gene Symbol: LOC689770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142304.1 | NP_001135776.1 | 632 | |||
GenBank mRNA | EU128747.1 | - | 638 | |||
FJ404474.1 | - | 632 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
