siRNA Details
Gene Symbol: Clec2dl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085404.1 | NP_001078873.1 | 4 | 556 | ||
XM_006237453.2 | XP_006237515.1 | 5 | 589 | |||
XM_008763333.1 | XP_008761555.1 | 4 | 1615 | |||
GenBank mRNA | EF100689.1 | - | 4 | 556 | ||
FJ404471.1 | - | 4 | 556 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
