siRNA Details
Gene Symbol: Clec2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177689.1 | NP_001171160.1 | 4 | 445 | ||
XM_008763336.1 | XP_008761558.1 | 5 | 2537 | |||
GenBank mRNA | FJ404473.1 | - | 4 | 481 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
