siRNA Details
Gene Symbol: Magea4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109550.1 | NP_001103020.1 | 2 | 292 | ||
XM_008773588.1 | XP_008771810.1 | 6 | 631 | |||
XM_008773589.1 | XP_008771811.1 | 6 | 641 | |||
XM_008773590.1 | XP_008771812.1 | 7 | 847 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
