siRNA Details
Gene Symbol: Smim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163002.1 | NP_001156474.1 | 3 | 852 | ||
NM_001277479.1 | NP_001264408.1 | 4 | 1010 | |||
XM_006247731.1 | XP_006247793.1 | 2 | 7073 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
