siRNA Details
Gene Symbol: LOC689991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767349.1 | XP_008765571.1 | 54 | 8119 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175510, FJ210934, AK173273.
