siRNA Details
Gene Symbol: Bag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128195.1 | NP_001121667.1 | 3 | 702 | ||
GenBank mRNA | BC158711.1 | - | 3 | 702 | 9035120 | |
FQ228417.1 | - | 3 | 726 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
